3-[(3-Methylbenzyl)oxy]piperidine

Supplier: Matrix Scientific

023162-500MG
101815-460EA 235.75 USD
101815-460
3-[(3-Methylbenzyl)oxy]piperidine
3-[(3-Methylbenzyl)oxy]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08688172
CAS Number: 946680-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR