3-[(3-Chlorobenzyl)oxy]piperidine hydrochloride
Supplier: Matrix Scientific
023161-500MG
101815-458EA
435.38
USD
101815-458
3-[(3-Chlorobenzyl)oxy]piperidine hydrochloride
3-[(3-Chlorobenzyl)oxy]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇Cl₂NO |
MDL Number:
MFCD12405040 CAS Number: 1185301-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...