3-[(3-Fluorobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023160-500MG
101815-456EA 435.38 USD
101815-456
3-[(3-Fluorobenzyl)oxy]piperidine hydrochloride
3-[(3-Fluorobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClFNO
MDL Number: MFCD11506390
CAS Number: 1184997-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR