3-[(2-Methylbenzyl)oxy]piperidine

Supplier: Matrix Scientific

023159-500MG
101815-454EA 315.38 USD
101815-454
3-[(2-Methylbenzyl)oxy]piperidine
3-[(2-Methylbenzyl)oxy]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD08688171
CAS Number: 946714-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR