3-(Benzyloxy)piperidine hydrochloride

Supplier: Matrix Scientific

023157-500MG
101815-450EA 495.38 USD
101815-450
3-(Benzyloxy)piperidine hydrochloride
3-(Benzyloxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD07781093
CAS Number: 37098-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR