3-Ethoxypiperidine hydrochloride

Supplier: Matrix Scientific

023155-500MG
101815-446EA 40.25 USD
101815-446
3-Ethoxypiperidine hydrochloride
3-Ethoxypiperidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO
MDL Number: MFCD09475416
CAS Number: 1159826-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR