3-(3-Phenylpropoxy)piperidine hydrochloride
Supplier: Matrix Scientific
023153-500MG
101815-442EA
495.38
USD
101815-442
3-(3-Phenylpropoxy)piperidine hydrochloride
3-(3-Phenylpropoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₂ClNO |
MDL Number:
MFCD09997063 CAS Number: 1185297-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...