3-[(3-Methyl-2-butenyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

023150-500MG
101815-436EA 538.46 USD
101815-436
3-[(3-Methyl-2-butenyl)oxy]piperidine hydrochloride
3-[(3-Methyl-2-butenyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MDL Number: MFCD12405038
CAS Number: 1185304-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR