3-(2-Methoxyethoxy)piperidine hydrochloride

Supplier: Matrix Scientific

023148-500MG
101815-432EA 538.46 USD
101815-432
3-(2-Methoxyethoxy)piperidine hydrochloride
3-(2-Methoxyethoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₈H₁₈ClNO₂
MDL Number: MFCD09997923
CAS Number: 1185054-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR