4-Chloro-2-fluorophenyl-4-piperidinylmethyl ether
Supplier: Matrix Scientific
023143-500MG
101815-424EA
315.38
USD
101815-424
4-Chloro-2-fluorophenyl-4-piperidinylmethyl ether
4-Chloro-2-fluorophenyl-4-piperidinylmethyl ether
Hazard: Irritant
Formula:
C₁₂H₁₅ClFNO MW: 243.71 g/mol |
MDL Number:
MFCD08687336 CAS Number: 946680-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...