2-Benzyl-4-chlorophenyl-4-piperidinylmethyl ether
Supplier: Matrix Scientific
023139-500MG
101815-416EA
315.38
USD
101815-416
2-Benzyl-4-chlorophenyl-4-piperidinylmethyl ether
2-Benzyl-4-chlorophenyl-4-piperidinylmethyl ether
Hazard: Irritant
Formula:
C₁₉H₂₂ClNO |
MDL Number:
MFCD08686857 CAS Number: 946680-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...