4-[(3-Ethoxyphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023136-500MG
101815-410EA 315.38 USD
101815-410
4-[(3-Ethoxyphenoxy)methyl]piperidine
4-[(3-Ethoxyphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD08687739
CAS Number: 946725-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR