4-[(3-Nitrophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023130-500MG
101815-400EA 315.38 USD
101815-400
4-[(3-Nitrophenoxy)methyl]piperidine
4-[(3-Nitrophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD08688447
CAS Number: 946725-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR