8-(4-Piperidinylmethoxy)quinoline dihydrochloride
Supplier: Matrix Scientific
023128-500MG
101815-396EA
402.5
USD
101815-396
8-(4-Piperidinylmethoxy)quinoline dihydrochloride
8-(4-Piperidinylmethoxy)quinoline dihydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₀Cl₂N₂O |
MDL Number:
MFCD12405035 CAS Number: 1185298-73-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...