2-Chloro-3-(4-piperidinylmethoxy)pyridine hydrochloride
Supplier: Matrix Scientific
023125-500MG
101815-390EA
538.46
USD
101815-390
2-Chloro-3-(4-piperidinylmethoxy)pyridine hydrochloride
2-Chloro-3-(4-piperidinylmethoxy)pyridine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆Cl₂N₂O |
MDL Number:
MFCD11506388 CAS Number: 1185298-34-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...