4-[({1-[(E)-2-Phenyldiazenyl]-2-naphthyl}oxy)methyl]piperidine

Supplier: Matrix Scientific

023121-500MG
101815-382EA 235.75 USD
101815-382
4-[({1-[(E)-2-Phenyldiazenyl]-2-naphthyl}oxy)methyl]piperidine
4-[({1-[(E)-2-Phenyldiazenyl]-2-naphthyl}oxy)methyl]piperidine

Hazard: Irritant

Formula: C₂₂H₂₃N₃O
MDL Number: MFCD08688477
CAS Number: 946758-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR