Methyl-2-[4-(4-piperidinylmethoxy)phenyl]acetate hydrochloride
Supplier: Matrix Scientific
023117-500MG
101815-374EA
315.38
USD
101815-374
Methyl-2-[4-(4-piperidinylmethoxy)phenyl]acetate hydrochloride
Methyl-2-[4-(4-piperidinylmethoxy)phenyl]acetate hydrochloride
Hazard: Irritant
Formula:
C₁₅H₂₂ClNO₃ |
MDL Number:
MFCD09879204 CAS Number: 1185153-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...