4-{[4-(sec-Butyl)phenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

023115-500MG
101815-370EA 920 USD
101815-370
4-{[4-(sec-Butyl)phenoxy]methyl}piperidine hydrochloride
4-{[4-(sec-Butyl)phenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD11100491
CAS Number: 1185301-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR