4-[(4-Phenoxyphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023108-500MG
101815-356EA 315.38 USD
101815-356
4-[(4-Phenoxyphenoxy)methyl]piperidine
4-[(4-Phenoxyphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₈H₂₁NO₂
MDL Number: MFCD08688468
CAS Number: 946713-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR