4-[(3,5-Dimethylphenoxy)methyl]piperidine
023103-500MG
101815-346EA
825.7
USD
101815-346
4-[(3,5-Dimethylphenoxy)methyl]piperidine
4-[(3,5-Dimethylphenoxy)methyl]piperidine
Hazard: Irritant
Formula:
C₁₄H₂₁NO |
MDL Number:
MFCD08687654 CAS Number: 946725-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...