4-[(3-Isopropylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

023101-500MG
101815-344EA 315.38 USD
101815-344
4-[(3-Isopropylphenoxy)methyl]piperidine hydrochloride
4-[(3-Isopropylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄ClNO
MDL Number: MFCD10687216
CAS Number: 1185165-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR