Methyl-3-(4-piperidinylmethoxy)benzoate hydrochloride

Supplier: Matrix Scientific

023099-500MG
101815-340EA 315.38 USD
101815-340
Methyl-3-(4-piperidinylmethoxy)benzoate hydrochloride
Methyl-3-(4-piperidinylmethoxy)benzoate hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₀ClNO₃
MDL Number: MFCD09879200
CAS Number: 1184976-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR