4-[(3-Bromophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

023091-500MG
101815-328EA 538.46 USD
101815-328
4-[(3-Bromophenoxy)methyl]piperidine hydrochloride
4-[(3-Bromophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD09997939
CAS Number: 1185301-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR