4-[(2-Ethylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023090-500MG
101815-326EA 315.38 USD
101815-326
4-[(2-Ethylphenoxy)methyl]piperidine
4-[(2-Ethylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD08687780
CAS Number: 946713-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR