4-[(2-Methylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023086-500MG
101815-318EA 315.38 USD
101815-318
4-[(2-Methylphenoxy)methyl]piperidine
4-[(2-Methylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD06248329
CAS Number: 63608-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR