4-[([1,1'-Biphenyl]-2-yloxy)methyl]piperidine

Supplier: Matrix Scientific

023085-500MG
101815-316EA 315.38 USD
101815-316
4-[([1,1'-Biphenyl]-2-yloxy)methyl]piperidine
4-[([1,1'-Biphenyl]-2-yloxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₈H₂₁NO
MDL Number: MFCD06245983
CAS Number: 883514-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR