2-Chloro-5-methylphenyl-4-piperidinylmethyl ether

023082-500MG
101815-310EA 235.75 USD
101815-310
2-Chloro-5-methylphenyl-4-piperidinylmethyl ether
2-Chloro-5-methylphenyl-4-piperidinylmethyl ether

Hazard: Irritant

Formula: C₁₃H₁₈ClNO
MDL Number: MFCD06246354
CAS Number: 883517-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR