4-[(2,5-Dichlorophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023081-500MG
101815-308EA 315.38 USD
101815-308
4-[(2,5-Dichlorophenoxy)methyl]piperidine
4-[(2,5-Dichlorophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂NO
MDL Number: MFCD08687536
CAS Number: 946680-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR