4-[(2,4-Dichlorophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023080-500MG
101815-306EA 315.38 USD
101815-306
4-[(2,4-Dichlorophenoxy)methyl]piperidine
4-[(2,4-Dichlorophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂NO
MDL Number: MFCD08687535
CAS Number: 946713-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR