4-[(2-Chlorophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023078-500MG
101815-302EA 315.38 USD
101815-302
4-[(2-Chlorophenoxy)methyl]piperidine
4-[(2-Chlorophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₆ClNO
MDL Number: MFCD06246403
CAS Number: 63608-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR