4-[(2-Bromophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

023075-500MG
101815-298EA 235.75 USD
101815-298
4-[(2-Bromophenoxy)methyl]piperidine hydrochloride
4-[(2-Bromophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇BrClNO
MDL Number: MFCD09997061
CAS Number: 1185297-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR