4-(Phenoxymethyl)piperidine hydrochloride

Supplier: Matrix Scientific

023074-500MG
101815-296EA 461.54 USD
101815-296
4-(Phenoxymethyl)piperidine hydrochloride
4-(Phenoxymethyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD09879196
CAS Number: 63608-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR