4-[(5,6,7,8-Tetrahydro-1-naphthalenyloxy)methyl]piperidine

Supplier: Matrix Scientific

023073-500MG
101815-294EA 315.38 USD
101815-294
4-[(5,6,7,8-Tetrahydro-1-naphthalenyloxy)methyl]piperidine
4-[(5,6,7,8-Tetrahydro-1-naphthalenyloxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₆H₂₃NO
MDL Number: MFCD06248814
CAS Number: 883539-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR