4-[2-(4-Piperidinylmethoxy)ethyl]morpholine

Supplier: Matrix Scientific

023072-500MG
101815-292EA 315.38 USD
101815-292
4-[2-(4-Piperidinylmethoxy)ethyl]morpholine
4-[2-(4-Piperidinylmethoxy)ethyl]morpholine

Hazard: Irritant

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD08688497
CAS Number: 946680-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR