2-Methyl-2-propenyl-4-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

023071-500MG
101815-290EA 538.46 USD
101815-290
2-Methyl-2-propenyl-4-piperidinylmethyl ether hydrochloride
2-Methyl-2-propenyl-4-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO
MDL Number: MFCD11100490
CAS Number: 1185297-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR