4-{[(4-Methoxyphenethyl)oxy]methyl}piperidine

Supplier: Matrix Scientific

023055-500MG
101815-260EA 235.75 USD
101815-260
4-{[(4-Methoxyphenethyl)oxy]methyl}piperidine
4-{[(4-Methoxyphenethyl)oxy]methyl}piperidine

Hazard: Irritant

Formula: C₁₅H₂₃NO₂
MDL Number: MFCD08688068
CAS Number: 933701-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR