4-[(2-Ethoxyethoxy)methyl]piperidine

Supplier: Matrix Scientific

023054-500MG
101815-258EA 235.75 USD
101815-258
4-[(2-Ethoxyethoxy)methyl]piperidine
4-[(2-Ethoxyethoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₀H₂₁NO₂
MDL Number: MFCD08687700
CAS Number: 265108-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR