4-Chloro-2-fluorophenyl-3-piperidinylmethyl ether

Supplier: Matrix Scientific

023051-500MG
101815-252EA 235.75 USD
101815-252
4-Chloro-2-fluorophenyl-3-piperidinylmethyl ether
4-Chloro-2-fluorophenyl-3-piperidinylmethyl ether

Hazard: Irritant

Formula: C₁₂H₁₅ClFNO
MDL Number: MFCD08687335
CAS Number: 946680-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR