3-[(2,4-Dichloro-3,5-dimethylphenoxy)methyl]piperidine

Supplier: Matrix Scientific

023046-500MG
101815-242EA 315.38 USD
101815-242
3-[(2,4-Dichloro-3,5-dimethylphenoxy)methyl]piperidine
3-[(2,4-Dichloro-3,5-dimethylphenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₄H₁₉Cl₂NO
MW: 288.22 g/mol
MDL Number: MFCD08687526
CAS Number: 946680-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR