2-Methoxy-5-nitrophenyl-3-piperidinylmethyl ether

Supplier: Matrix Scientific

023043-500MG
101815-236EA 315.38 USD
101815-236
2-Methoxy-5-nitrophenyl-3-piperidinylmethyl ether
2-Methoxy-5-nitrophenyl-3-piperidinylmethyl ether

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄
MDL Number: MFCD08688065
CAS Number: 946680-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR