3-[(4-Bromo-2-fluorophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

023042-500MG
101815-234EA 315.38 USD
101815-234
3-[(4-Bromo-2-fluorophenoxy)methyl]piperidine hydrochloride
3-[(4-Bromo-2-fluorophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆BrClFNO
MDL Number: MFCD11100489
CAS Number: 1185298-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR