3-[(2,4-Difluorophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023040-500MG
101815-230EA 315.38 USD
101815-230
3-[(2,4-Difluorophenoxy)methyl]piperidine
3-[(2,4-Difluorophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₅F₂NO
MDL Number: MFCD08687566
CAS Number: 946725-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR