3-[(3-Methyl-4-nitrophenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
023039-500MG
101815-228EA
315.38
USD
101815-228
3-[(3-Methyl-4-nitrophenoxy)methyl]piperidine hydrochloride
3-[(3-Methyl-4-nitrophenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₃ |
MDL Number:
MFCD11100488 CAS Number: 1185301-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...