3-[(3-Nitrophenoxy)methyl]piperidine

Supplier: Matrix Scientific

023038-500MG
101815-226EA 315.38 USD
101815-226
3-[(3-Nitrophenoxy)methyl]piperidine
3-[(3-Nitrophenoxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD08688446
CAS Number: 946713-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR