7-(3-Piperidinylmethoxy)-2H-chromen-2-one

Supplier: Matrix Scientific

023037-500MG
101815-224EA 315.38 USD
101815-224
7-(3-Piperidinylmethoxy)-2H-chromen-2-one
7-(3-Piperidinylmethoxy)-2H-chromen-2-one

Hazard: Irritant

Formula: C₁₅H₁₇NO₃
MDL Number: MFCD08688494
CAS Number: 946724-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR