8-(3-Piperidinylmethoxy)quinoline

Supplier: Matrix Scientific

023036-500MG
101815-222EA 315.38 USD
101815-222
8-(3-Piperidinylmethoxy)quinoline
8-(3-Piperidinylmethoxy)quinoline

Hazard: Irritant

Formula: C₁₅H₁₈N₂O
MDL Number: MFCD08688504
CAS Number: 946680-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR