2-Methyl-5-(3-piperidinylmethoxy)pyridine

Supplier: Matrix Scientific

023034-500MG
101815-218EA 235.75 USD
101815-218
2-Methyl-5-(3-piperidinylmethoxy)pyridine
2-Methyl-5-(3-piperidinylmethoxy)pyridine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD08688364
CAS Number: 914299-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR