2-Chloro-3-(3-piperidinylmethoxy)pyridine

Supplier: Matrix Scientific

023033-500MG
101815-216EA 235.75 USD
101815-216
2-Chloro-3-(3-piperidinylmethoxy)pyridine
2-Chloro-3-(3-piperidinylmethoxy)pyridine

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD08687467
CAS Number: 946713-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR