3-[(2-Naphthyloxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

023031-500MG
101815-212EA 315.38 USD
101815-212
3-[(2-Naphthyloxy)methyl]piperidine hydrochloride
3-[(2-Naphthyloxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClNO
MDL Number: MFCD28023812
CAS Number: 1706435-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR