3-[(1-Naphthyloxy)methyl]piperidine

Supplier: Matrix Scientific

023030-500MG
101815-210EA 315.38 USD
101815-210
3-[(1-Naphthyloxy)methyl]piperidine
3-[(1-Naphthyloxy)methyl]piperidine

Hazard: Irritant

Formula: C₁₆H₁₉NO
MDL Number: MFCD06248554
CAS Number: 738521-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR